Product Image
Thermo Scientific™
UltiMate™ 3000 Autosampler Bridges
Order Thermo Scientific™ Dionex™ UltiMate™ 3000 Autosampler Bridges when you need replacements. They are available in 1μL (PEEKsil) and 6μL (PEEK) volumes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Sample Capacity
6825.0015WPS-3000 TBFC Analytical for Nano/Cap1 μL
6825.0016WPS-3000 TBFC Analytical6.2 μL
6824.0015WPS-3000 TFC Analytical for Nano/Cap1 μL
6824.0016WPS-3000 TFC Analytical6.2 μL
Catalog number 6825.0015
Price (INR)
-
For Use With (Equipment):
WPS-3000 TBFC Analytical for Nano/Cap
Sample Capacity:
1 μL
Order Thermo Scientific™ Dionex™ UltiMate™ 3000 Autosampler Bridges when you need replacements. They are available in 1μL (PEEKsil) and 6μL (PEEK) volumes.
Specifications
For Use With (Equipment)WPS-3000 TBFC Analytical for Nano/Cap
MaterialPEEKsil
Sample Capacity1 μL
TypeBridge
Unit SizeEach