Thermo Scientific™
Sample Loops for UltiMate™ WPS-3000T PL RS Pulled Loop Autosamplers
Catalog number: 6826.2401
Thermo Scientific™
Sample Loops for UltiMate™ WPS-3000T PL RS Pulled Loop Autosamplers
Catalog number: 6826.2401
Order the Thermo Scientific™ Dionex™ Sample Loops (1-125µL) you need for the Thermo Scientific™ Dionex™ UltiMate™ WPS-3000T PL RS pulled loop autosampler. The sample loops incorporate Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings which eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system every time.
Catalog Number
6826.2401
Unit Size
Each
For Use With (Equipment)
Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Sample Capacity
1 μL
Price (USD)
Full specifications
TypeSample Loop, NanoViper
Sample Capacity1 μL
Max. Pressure1000 bar
For Use With (Equipment)Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Sample Capacity
5 μL
Price (USD)
Full specifications
TypeSample Loop, NanoViper
Sample Capacity5 μL
Max. Pressure1000 bar
For Use With (Equipment)Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Sample Capacity
20 μL
Price (USD)
Full specifications
TypeSample Loop, NanoViper
Sample Capacity20 μL
Max. Pressure1000 bar
For Use With (Equipment)Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Sample Capacity
50 μL
Price (USD)
Full specifications
TypeSample Loop, NanoViper
Sample Capacity50 μL
Max. Pressure1000 bar
For Use With (Equipment)Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Sample Capacity
125 μL
Price (USD)
Full specifications
TypeSample Loop, NanoViper
Sample Capacity125 μL
Max. Pressure1000 bar
For Use With (Equipment)Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Unit SizeEach
Catalog Number
6826.2410
also known as 6826.241
Unit Size
Each
For Use With (Equipment)
Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Sample Capacity
10 μL
Price (USD)
Full specifications
TypeSample Loop, NanoViper
Sample Capacity10 μL
Max. Pressure1000 bar
For Use With (Equipment)Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeFor Use With (Equipment)Sample CapacityPrice (USD)
6826.2401Full specifications
EachDionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS1 μLRequest A Quote
TypeSample Loop, NanoViper
Sample Capacity1 μL
Max. Pressure1000 bar
For Use With (Equipment)Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Unit SizeEach
6826.2405Full specifications
EachDionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS5 μLRequest A Quote
TypeSample Loop, NanoViper
Sample Capacity5 μL
Max. Pressure1000 bar
For Use With (Equipment)Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Unit SizeEach
6826.2420Full specifications
EachDionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS20 μLRequest A Quote
TypeSample Loop, NanoViper
Sample Capacity20 μL
Max. Pressure1000 bar
For Use With (Equipment)Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Unit SizeEach
6826.2450Full specifications
EachDionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS50 μLRequest A Quote
TypeSample Loop, NanoViper
Sample Capacity50 μL
Max. Pressure1000 bar
For Use With (Equipment)Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Unit SizeEach
6826.2412Full specifications
EachDionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS125 μLRequest A Quote
TypeSample Loop, NanoViper
Sample Capacity125 μL
Max. Pressure1000 bar
For Use With (Equipment)Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Unit SizeEach
6826.2410
also known as 6826.241
Full specifications
EachDionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS10 μLRequest A Quote
TypeSample Loop, NanoViper
Sample Capacity10 μL
Max. Pressure1000 bar
For Use With (Equipment)Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Unit SizeEach
Showing 6 of 6
nanoViper is the new, fingertight connection system for nano LC connections which eliminates the assembly of PEEK™ sleeve connections. It is preassembled to maximize ease-of-use, and is fingertight, making over tightening impossible. The nanoViper fitting is capable of withstanding pressures up to a 1000 bar and is compatible with third party valves and unions.