Search
| Catalog Number | Sample Capacity |
|---|---|
| 6826.2410 | 10 μL |
| 6826.2401 | 1 μL |
| 6826.2405 | 5 μL |
| 6826.2420 | 20 μL |
| 6826.2450 | 50 μL |
| 6826.2412 | 125 μL |
Order the Thermo Scientific™ Dionex™ Sample Loops (1-125μL) you need for the Thermo Scientific™ Dionex™ UltiMate™ WPS-3000T PL RS pulled loop autosampler. The sample loops incorporate Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings which eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system every time.