Product Image
Thermo Scientific™
Sample Loops for UltiMate™ ACC-3000T Column Compartment
Order the sample loop (20, 50, and 200μL) you need for the Thermo Scientific™ Dionex™ UltiMate™ ACC-3000T Column Compartment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Capacity
6830.241650 μL
6830.2418200 μL
6830.241420 μL
Catalog number 6830.2416
Price (MXN)
-
Sample Capacity:
50 μL
Order the Thermo Scientific™ Dionex™ Sample Loop (20, 50, and 200μL) you need for use in the Thermo Scientific™ Dionex™ UltiMate™ ACC-3000T Column Compartment. The sample loops incorporate Thermo Scientific™ Dionex™ Viper™ Fingertight Fittings which eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system every time.
See Viper and nanoViper resources
Specifications
For Use With (Equipment)UltiMate ACC-3000T Column Compartment
Sample Capacity50 μL
TypeSample Loop
Unit SizeEach