Vanquish™ Split Sampler Sample Loops
Actual product may vary
Thermo Scientific™
Vanquish™ Split Sampler Sample Loops
Thermo Scientific™ Vanquish™ Split Sampler Sample Loops come in a variety of sizes to fit your application requirements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Sample CapacityBiocompatible Version
6851.1970Vanquish Split Sampler C/CT250 μLNo
6850.1915Vanquish Split Sampler HT and FT10 μLYes
6850.1911Vanquish Split Sampler HT and FT25 μLYes
6850.1913Vanquish Split Sampler HT and FT100 μLYes
6851.1980Vanquish Split Sampler C/CT1000 μLNo
6851.1960Vanquish Split Sampler C/CT10 μLNo
6851.1950Vanquish Split Sampler C/CT100 μLNo
6851.1940Vanquish Split Sampler C/CT25 μLNo
6252.1940Vanquish Split Sampler NT25 μLYes
6252.1950Vanquish Split Sampler NT100 μLYes
6252.1960Vanquish Split Sampler NT10 μLYes
Catalog number 6851.1970
Price (MXN)
-
For Use With (Equipment):
Vanquish Split Sampler C/CT
Sample Capacity:
250 μL
Biocompatible Version:
No
Automate sample injection into your Thermo Scientific™ Vanquish™ HPLC or UHPLC system with Thermo Scientific™ Vanquish™ Split Sampler Sample Loops. These sample loops are available in 10, 25, 100, 250, 1000 μL sizes to meet your application requirements. Thermo Scientific™ Viper™ Fittings are included with our sample loops to make them easy to install with leak-free connections.
Vanquish Split Sampler Sample Loops use Viper Fittings, which have a ferrule-free tightening design, for virtually zero-dead-volume connections.
Specifications
Biocompatible VersionNo
For Use With (Equipment)Vanquish Split Sampler C/CT
MaterialStainless Steel
Sample Capacity250 μL
TypeSample Loop
Unit SizeEach