CD20 Monoclonal Antibody (2H7), eFluor™ 506, eBioscience™ - FAQs
CD20 Monoclonal Antibody (2H7), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD20 Monoclonal Antibody (2H7), eFluor™ 506, eBioscience™ - FAQs (69-0209-42, 69-0209-41)