Search
| Número do catálogo | Type |
|---|---|
| 6900.1020 | Well Plate and Sample Rack Shelf |
| 6900.1010 | Well Plate Shelf |
The 9-level shelf accommodates any 9 of the following (SBS footprint):
The 20-level shelf accommodates any 20 of the following (SBS footprint):
The optional Vanquish Charger module is an intelligent robotic that is fully integrated into the Thermo Scientific™ Vanquish™ UHPLC system. With a capacity of up to 8832 samples (23 well plates), the Charger module automates sample loading into the Vanquish UHPLC system for extra-long unattended operation. Environmental controls ensure reliable sample protection. A barcode reader automatically inventories each plate or rack within the device every time the door is closed, allowing users to add samples at anytime even while the Vanquish UHPLC system is in operation.