CaptureSelect™ KappaXP GoPure™ Column
Thermo Scientific™

CaptureSelect™ KappaXP GoPure™ Column

CaptureSelect KappaXP GoPure columns are ready-to-use columns designed for fast and easy purification of kappa monoclonal antibodies on standard chromatography systems with high purity and yield in a single step.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
69432120055 mL
69432120011 mL
Catalog number 6943212005
Price (CLP)
-
Quantity:
5 mL

The pre-packed column contains CaptureSelect KappaXP Affinity Matrix, which enables the purification of human immunoglobulins and Fab fragments containing the constant domain of the human kappa light chain.

Features of this affinity matrix include:

  • Single-step purification of immunogloblulins, Fab, and Fab2 fragments directly from complex source materials
  • Mild elution for Fab fragments and antibodies
  • Elution possible at pH 6.0 with 1.5 M MgCl2
  • Excellent pH stability up to 100 mM NaOH for limited exposure
  • Specific for human antibodies, works in the presence of serum (bovine/horse)
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

Matrix: agarose-based, epoxide-activated

Average particle size: 65 ± 10 μm

Ligand: CaptureSelect KappaXP affinity ligand

Fab binding capacity: 20–30 g IgG Kappa-Fab/liter resin, depending on residence time

IgG binding capacity: 35–45 g IgG/liter resin, depending on residence time

Elution conditions: 20 mM citric acid or acetic acid, pH 3–4; 100 mM Tris, 1.5 M MgCl2, pH 6

Column dimensions: 7 x 25 mm (1 mL column); 16 x 25 mm (5 mL column)

Max. recommended flow: 5 mL/min. (1 mL column); 10 mL/min. (5 mL column)

Max. operation pressure: 2 bar

Formulation buffer: 20% (v/v) ethanol

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bed Height25 mm
Bed Volume5 mL
Diameter (Metric)16 mm
MatrixKappaXP Affinity Matrix
Max. Operating Pressure0.2 MPa
Quantity5 mL
Capacity (Metric)Load capacity for 5 mL resin
FormAgarose Bead
Particle Size65 ±10 μm
Product LineCaptureSelect
TypePre-packed Column
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C in dark/protect from light.