Thermo Scientific™

CaptureSelect™ FcXP EvolveD™ Column

Catalog number:  6943712202

Back to top

Description

CaptureSelect FcXP EvolveD columns are ready-to-use columns designed specifically for use in biopharmaceutical applications. These pre-packed columns are suitable for cGMP manufacturing and have the following features:
• Prepared and packed in a classified ISO7 environment
• An entirely non-metallic flow path
• Qualified in terms of performance and pressure flow characteristics
• Suitable for direct connection to standard chromatography systems
• Available Regulatory Support File
• Packed with CaptureSelect FcXP Affinity Matrix, offering high dynamic binding capacity and high resolution over a wide range of flow rates

Please note that CaptureSelect EvolveD affinity columns are made to order. They are custom manufactured two-at-a-time, so are offered only in quantities that are a multiple of two. For detailed information on the resin, please see the CaptureSelect EvolveD column manual below.

Operating and technical specifications
Column type10-cm bed height20-cm bed height
Column diameter (cm)7102071020
Column length (cm)101010202020
Cross-sectional area (cm2)38.578.5314.238.578.5314.2
Column volume (mL)385785314277015706284

• Pressure rating (bar/psi): 4/58
• Operating temperature: 2–30°C, dependent on the resin packed
• Bed support size: 8 µm
• Inlet/outlet internal diameter: 3 mm or 6 mm depending on the column diameter
• Inlet/outlet port connections: 25-mm TC sanitary fitting

View our comprehensive column overview › (Select the Antibody Purification tab.)

CaptureSelect FcXP Affinity Matrix can be utilized for the purification of IgG and Fc fusion proteins directly from complex source materials and enables a robust and efficient purification process with excellent purity obtained in one step.

Features of this affinity matrix include:
• Mild elution, making it suitable for Fc fusion proteins
• Human specific, no binding to bovine antibodies
• Excellent scalability
• Non-animal derived

View other biosimilars and recombinant proteins affinity matrices ›

Free of animal components
CaptureSelect products are affinity ligands created with proprietary technology based on camelid-derived single-domain antibody fragments. The FcXP ligand is a 13-kDa fragment comprising the three complementarity determining regions (CDRs) that form the antigen binding domain. The ligand is efficiently produced by the yeast Saccharomyces cerevisiae in a production process free of animal components.

Main characteristics:

Matrix: agarose-based, epoxide-activated
Average particle size: 65 ± 10 µm
Ligand: CaptureSelect FcXP affinity ligand
Ligand coupling method: epoxide coupling
Binding capacity: 40–50 g IgG/L resin depending on flow rate, column height, and contact time
Elution conditions: 20 mM citric acid or acetic acid, pH 3–4
Max. operation pressure: 2 bar
Formulation buffer: 20% (v/v) ethanol

Specifications

Format: EvolveD Column
Capacity (Metric): 6284 mL
Product Type: Agarose bead
Quantity: 1 Pc.