Thermo Scientific™
Richard-Allan Scientific™ Signature Series Clear-Rite™ 3
Related applications:
Clinical
Render complete deparaffinization and clearing during the staining process with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Clear-Rite™ 3, a unique blend of isoparaffinic aliphatic hydrocarbons.
Back to top
  • Very low odor; Benzene-free
  • Use in all tissue processors and automatic stainers
  • Does not produce hard or brittle specimens, even after prolonged exposure