SureSTART™ Specification Certified 2 mL Short Thread Screw Amber Glass SureStop™ Vial and 9 mm AVCS™ Screw Cap Kits with White Silicone/Red PTFE Septa, Level 2 - FAQs
SureSTART™ Specification Certified 2 mL Short Thread Screw Amber Glass SureStop™ Vial and 9 mm AVCS™ Screw Cap Kits with White Silicone/Red PTFE Septa, Level 2 - FAQs
View additional product information for SureSTART™ Specification Certified Screw Vial and Cap Kits, Level 2 High-throughput Applications - FAQs (6AK78BW, 6AK94W, 6AK82W, 6AKSV9PP-15PI, 6AK92W, 6AK80W, 6AK86W, 6AK95W, 6AK594W, 6AK595W, 6AK188W, 6AK599W, 6AK93BW, 6AK195W, 6AK196W, 6AK186W, 6AK592W, 6AK180W, 6AK192W)
1 product FAQs found
What are SureStop vials?
In addition to the features on standard 2 mL vials, SureStop vials also have a patented definite stop point (alignment ring) in the design of the vial finish. This prevents the vial from being an optimal seal, which eliminates under- and over-sealing by the cap/closure and user-to-user variance.
SureStop vials are designed for use with Advanced Vial Closure System (AVCS) screw caps. They improve transfer reliability and give a more secure liquid seal than standard screw vials. SureStop vials are available as a 2 mL as either clear or amber options and are highly recommended for use with GC and LC applications.