SureSTART™ 1.7 mL Clear Glass High Recovery Screw Top Microvials, 5000/pack, Level 2 - FAQs
SureSTART™ 1.7 mL Clear Glass High Recovery Screw Top Microvials, 5000/pack, Level 2 - FAQs
View additional product information for SureSTART™ 1.7 mL Glass High Recovery Screw Top (U)HPLC and GC Vials, Level 2 - FAQs (6ASV9-V1F)
1 product FAQs found
What are SureSTART microvials?
Microvials are suitable for use when sample volume is limited or when you need to get maximum recovery from the vial. These vials allow small volumes to be sampled accurately by autosamplers with a recoverable volume of much less than 100 µL. High recovery, total recovery, and vials with fixed inserts are all classed as microvials.