Search
| Catalog Number | Quantity |
|---|---|
| 6PRV11-1P | 100/Pk. |
| 6PRV11-1PF | 5000/Pk. |
| 6PRV11-1PT | 1000/Pk. |
Thermo Scientific™ SureSTART™ 2 mL Snap Top Vials (Level 3) are the premier choice for high sensitivity LC-MS workflows, delivering reproducible results and exceptional sample protection for demanding applications.
Made with high purity clear glass, these vials contain the lowest metal content, extractables, and leachables – critical for meeting US, EU, and JPN Pharmacopeia standards. Also ensures consistent sensitivity and selectivity across HRAM (high resolution accurate mass) instruments including Thermo Scientific™ LC Orbitrap™ MS systems.
Combine with Thermo Scientific™ SureSTART™ 11 mm snap caps and septa, to ensure optimal performance.
Key Features
Performance Level 3 — for High-Sensitivity Applications
Specification Certified
Lot Traceable