Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
主页
▸
订购所有产品
▸
纯化系统
▸
液相色谱仪
▸
单独的 HPLC 部件和附件
▸
HPLC 检测器
▸
LC 电化学检测器在线过滤器
Thermo Scientific™
LC 电化学检测器在线过滤器
LC 电化学检测器内联过滤器,包括进样器前、后过滤器套件和过滤器替换装置。
Have Questions?
更改视图
货号
类型
材料
70-0893
在线颗粒过滤器套件
石墨
70-0898
在线颗粒过滤器套件
石墨
70-3824
在线颗粒过滤器套件
PEEK
70-4093
在线颗粒过滤器套件
PEEK
4 选项
货号
70-0893
价格(TWD)
-
申请报价
类型:
在线颗粒过滤器套件
材料:
石墨
使用 Thermo Scientific™ 电化学检测器内联过滤器,包括进样器前、后过滤器套件和过滤器替换装置。
规格
描述
进样器前过滤器套件,带石墨滤芯
适用于(应用)
Pre-injector Locations
适用于(设备)
Coulochem 和 CoulArray 检测器系列
材料
石墨
数量
1 Ea.
检测器类型
电化学的
类型
在线颗粒过滤器套件