LC Electrochemical Detectors In-Line Filters
LC Electrochemical Detectors In-Line Filters
Actual product may vary
Thermo Scientific™
LC Electrochemical Detectors In-Line Filters
LC Electrochemical Detectors In-Line Filters which include pre- and post-injector filter kits and replacement filter units.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeMaterial
70-3824Filter ElementsPEEK
70-0893Filter KitGraphite
70-0898Filter ElementsGraphite
70-4093Filter KitPEEK
Catalog number 70-3824
Price (USD)
-
Request A Quote
Type:
Filter Elements
Material:
PEEK
Use Thermo Scientific™ LC Electrochemical Detectors In-Line Filters that include pre- and post-injector filter kits and replacement filter units.
Specifications
For Use With (Application)for Post-injector locations
For Use With (Equipment)Coulochem and CoulArray Detector Series
MaterialPEEK
Quantity5/Pk.
Detector TypeElectrochemical (ECD)
TypeFilter Elements
Unit SizeEach