Versa Chain Checkweigher
Versa Chain Checkweigher
Versa Chain Checkweigher
Versa Chain Checkweigher
Versa Chain Checkweigher
Versa Chain Checkweigher
Versa Chain Checkweigher
Thermo Scientific™

Versa Chain Checkweigher

Improve throughput efficiencies and increase your bottom line with the Thermo Scientific™ Versa Chain Checkweigher.
Have Questions?
Catalog NumberDescription
70.181Versa Chain Checkweigher
Catalog number 70.181
Price (USD)
-
Request A Quote
Description:
Versa Chain Checkweigher

Improve throughput efficiencies and increase your bottom line with the Thermo Scientific™ Versa Chain Checkweigher. With line speeds up to 700 packages per minute, the Versa Chain Checkweigher provides reliable high-speed weighing, control and reject functions for cartons, cans, bottles and pouches in dry or wet environments.

The Versa Chain Checkweigher Features:

  • Open chain design to minimize product build-up and facilitate easy cleaning and maintenance
  • Durable weighframe with low-maintenance drive system
  • Configurable security access levels
  • One hundred standard product memories
  • Easy-to-read interface with touch-screen operation and high-visibility weight display
  • Stand-alone pedestal or with standard integrated conveyor
  • Optional infeed conveyor fitted with peripheral equipment
  • Optional metal detector can be added for complete weight control and contaminant inspection
  • OIML MID Approved

Ordering Alerts
Upgrade existing frames to new controller: A conversion kit available to upgrade checkweigher's electronics to the Versa controller if existing frame is in good condition.

Recommended for: Food, beverage, pharmaceutical and consumer goods industries

Specifications
Accuracy±0.5 g, micropitch chain; ±1.0 g, nylatrin chain; ±1.5 g, stainless-steel chain (product/speed dependent); 2 standard deviations from mean
DescriptionVersa Chain Checkweigher
Electrical Requirements115/220VAC +10% 50/60 Hz
Frame304 stainless steel
Frequency50/60 Hz
Height (English) Line26 - 42 in., adjustable in 2-in. increments
Height (Metric) Line660.4 - 1,066.8 mm, adjustable in 50.8-mm increments
Operating Temperature-10°C to +40°C (14°F to 104°F); 0-95% humidity
Product TypeVersa Chain Checkweigher
Speed Range (English)50 - 550 ft. per minute
Speed Range (Metric)6.1 - 167.6 m per minute
Voltage115/220VAC +10%
Weight (English) Max.Up to 25 lb. per product
Weight (Metric) Max.Up to 11 kg per product
Display TypeTouchscreen
Package TypeCartons, Cans, Bottles, and Pouches
Washdown OptionYes
Unit SizeEach

Frequently asked questions (FAQs)

How fast can machines check the weight of pet food products?

For pet food products in cartons, cans, bottles, or pouches, a chain checkweigher offers line speeds up to 700 packages per minute. It is appropriate for both dry and wet environments.

How can I tell if final packaged pet food products are being under or overfilled?

To ensure that pet food processors are not under or overfilling packages, checkweighers are used to verify the correct weight of each package. Accurate weight avoids giving away costly product or from receiving hefty fines from regulatory bodies for underfilling.

Do checkweighers also monitor for physical contaminants?

Checkweighers do not detect physical contaminants but they can be combined with food metal detectors and X-ray inspection systems that do detect contaminants. Food metal detection systems provide reliable, cost-effective protection from even the smallest metal contaminants found anywhere in a food production process. Food X-ray inspection systems provide protection from metal, glass, stone and other dense foreign objects for almost any type of packaged, bulk, or piped product.

Food manufacturers can find metallic and non-metallic foreign objects and eliminate “wet” product effects common with metal detectors with the Thermo Scientific NextGuard X-ray Detection Systems (https://www.thermofisher.com/order/catalog/product/NEXTGUARDPRO). Designed for a wide variety of food applications, NextGuard systems offer enhanced capabilities to inspect packaged products for missing pieces or components, under and over-fills, and other quality problems.

Can checkweighers be washed down?

Checkweighers can sometimes come in contact with food products that can leak or spill. The equipment has to get washed down regularly to prevent contamination so it must meet IP65 Washdown requirements. Therefore, checkweigher components require materials that can withstand washdown using chemicals. The metal should be fully constructed of ANSI Type 304 SS (stainless steel), which is an austenite steel. For even harsher situations, ANSI 316 stainless is used; it is more impervious to corrosion than 304 SS. Equipment should meet American National Standard ANSI/IEC 60529-2004 for equipment with electrical components. This standard describes a system for classifying the degrees of protection provided by enclosures of electrical equipment for two conditions:

- The protection of persons against access to hazardous parts and protection of equipment against the ingress of solid foreign objects
- The ingress of water

Equipment should be designed with minimal flat, horizontal surfaces and slots to reduce food buildup, and use stainless steel conveyor beds. IP65 enclosures provide basic waterproof protection from water, including jet sprays used during washdown.

What factors influence weighing accuracy using checkweighers?

Proper design of the in-line checkweigher can influence accuracy. Here are some of the design factors:

- Product speed
- Product weight and size
- Product transfer onto and off of the checkweigher
- Reject method and design

In addition, conveyors must be level and aligned, belts and chains should be inspected regularly for wear or product buildup, gaps between conveyors should be minimized and infeed and weigh table belts or chains should be running at the same speeds. Other factors include the spacing between products' centers to ensure that more than one product is not on the weighing table at any one time, and calibrations should be done regularly.