Thermo Scientific™
Combination Checkweigher and Metal Detector
Eliminate the expense of two machines with the Thermo Scientific™ Combination Checkweigher and Metal Detector.  Combining the superior performance of a Thermo Scientific™ VersaWeigh Checkweigher with the high sensitivity of a Thermo Scientific™ APEX Metal Detector, the Combination Checkweigher and Metal Detector gives you both accurate weight control and the assurance that your product is free of metal contaminants—all in a single space-saving instrument.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
The Combination Checkweigher and Metal Detector Features:
  • Separation of metal-contaminated products from potentially reusable off-weight products
  • Independent reject operation from the metal detector and checkweigher output signals
  • Basic or advanced level checkweigher to fit application
  • Highest level of sensitivity and stability with the lowest levels of false rejects in metal detector
Advantages:
  • One vendor for both product inspection requirements
  • One service technician to support both devices
  • Reduced number of product transfer points, line space requirements
  • Reduced
  • Reduced cost
Ordering Information
All Thermo Scientific™ checkweighers can be supplied in a combo configuration except the Thermo Scientific™ Teorema high-rate can weigher.
Recommended for: Food, beverage, pharmaceutical and consumer goods industries
Checkweigher comparison chart
Compare and find the checkweigher that’s right for you.
View checkweighers 
Documents
Manuals & protocols