These products are not currently available for ordering on ThermoFisher.com. Please continue to place orders through your existing ordering channel. As part of the transition to Thermo Fisher Scientific, these products will become available through Thermo Fisher ordering channels in the coming months.
LifeASSURE™ PLA Series Filter Cartridge
Thermo Scientific™
LifeASSURE™ PLA Series Filter Cartridge
LifeASSURE™ PLA Filter Cartridges combine a multi‐zone microporous membrane with Advanced Pleat Technology to optimize the usable surface area which may extend capacity. In a range of sizes for use as a prefilter or for bioburden control in pharmaceutical, biological and bioprocess applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModel
7000051579PLA045B03BX
7000050339PLA080B01BA
7100040348PLA020B02BB
7000029193PLA045B01BA
7000029197PLA065B01BA
7000029214PLA045B02BA
7000029215PLA045B03BA
7000050276PLA020B02BA
7000050328PLA020B01BK
7000050334PLA045B01CA
7000050924PLA020B03BK
7000051372PLA020B02BK
7100007246PLA020B03BA
7100043276PLA020B02CA
7100066079PLA020B01BA
7100066460PLA020B03BC
Catalog number 7000051579
Price (EUR)
-
Model:
PLA045B03BX
LifeASSURE™ PLA filter cartridges use multi‐zone microporous membranes with an open upstream zone that prefilters larger particles and a tighter downstream zone for high retention and contaminant capacity. Advanced Pleat Technology increases usable surface area and maintains open flow paths for longer service life. Used to reduce bioburden in buffers, media, and biological solutions. Durable and efficient, they deliver consistent performance and available in various sizes and end fittings.
  • Multi-zone microporous nylon membrane offers maximum membrane protection and increased contaminant capacity
  • Advanced Pleat Technology construction allows maximum fluid and contaminant access to filter surface area for highest throughput
  • 100% Integrity tested prior to release ensures consistent performance to specification
Specifications
Connectors226 O-ring and Spear
Diameter (English)2.76 in.
Diameter (Metric)7 cm
Filtration Area3.2 m²
For Use With (Application)Sterile Filtration, Biopharmaceutical Manufacturing
GradePLA045
Length (English)30 in.
Length (Metric)76.2 cm
ModelPLA045B03BX
Outlet FittingSingle Open End SOE
Outlet StyleSingle Open End SOE
Product LinePLA
Quantity6/Cs.
Temperature (English) Max.176°F
Temperature (Metric) Max80°C
TypeFilter Cartridge
Filter TypeMembrane Filter
Material (Membrane)Nylon 66 Multizone Microporous Membrane
Unit SizeCase of 6