Interested in a bulk or custom version of this product? Request pricing.

IL2 Recombinant Canine Protein

Catalog number: 70014DNAE50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Interleukin 2 (IL2), recombinant Canine Interleukin-2 / IL-2 protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the mature form of canine IL2 (Q29416) (Ala 21-Thr 155, 147Cys/Ser) fused to an initial Met at the N-terminus.

Activity:
1. Measured by its binding ability in a functional ELISA.
2. Immobilized recombinant Canine IL2 (cat:70014-DNAE) at 10 µg/ml can bind recombinant human IL2Ra-Fc (Cat:10165-H02H) with a linear range of 0.3125-20 µg/ml.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Oncology
Endotoxin Level: < 1 EU/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: IL2
Gene ID: 403989
Gene Symbol: IL2
Molecular Weight: 15.6 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Cytokines & Receptors
Protein Form: Full Length, Recombinant
Protein Name: Interleukin 2
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Dog⁄Canine
Tag Position: None
Validated Application: Assay Standard

Contents & storage

Includes 1 vials of protein in suspension buffer. Store at -20 °C..

Documents

Manuals & protocols

Recommended products