Nalgene™ Graduated HDPE Bucket
Nalgene™ Graduated HDPE Bucket
Nalgene™ Graduated HDPE Bucket
Nalgene™ Graduated HDPE Bucket
Thermo Scientific™

Nalgene™ Graduated HDPE Bucket

Transfer laboratory solutions and powders with the Thermo Scientific™ Nalgene™ Graduated HDPE Bucket, featuring a wide-lipped pour spout and polyethylene-coated handle.
製品番号(カタログ番号)容量(メートル法)
7002-0025PK
または、製品番号7002-0025
9.5 L
製品番号(カタログ番号) 7002-0025PK
または、製品番号7002-0025
価格(JPY)
9,660
キャンペーン価格
Ends: 26-Jun-2026
12,060
割引額 2,400 (20%)
Each
容量(メートル法):
9.5 L

Transfer laboratory solutions and powders with the Thermo Scientific™ Nalgene™ Graduated HDPE Bucket, featuring a wide-lipped pour spout and polyethylene-coated handle. This heavy-duty, rugged high density polyethylene bucket is chemically compatible with a wide range of lab reagents.

  • High-density polyethylene construction offers more durability than hard rubber
  • Polyethylene-coated wire bail handle provides a comfortable and sturdy grip
  • Molded-in graduations in pints and liters are easy to read
Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 

 

 

仕様
容量(ヤードポンド法)2.5 gal.
概要2.5 gal. (9.5 L) Bucket
ハンドル材質Polyethylene Coated
内容Handle
数量1 Ea.
形状Wide-Lipped Pour Spout
容量(メートル法)9.5 L
目盛りりMolded
材料HDPE
Unit SizeEach