RNase AWAY™ Surface Decontaminant - FAQs
RNase AWAY™ Surface Decontaminant - FAQs
View additional product information for RNase AWAY™ Surface Decontaminant - FAQs (7002, 7003PK, 7002PK, 7005-11, 7003, 7000TS)
1 product FAQs found
Do you have a Technical data sheet (TDS) for RNase AWAY Surface Decontaminant?
We have a safety data sheet (SDS) and COA for RNase AWAY Surface Decontaminant. Visit our Documents and Certificates page to search for and request these documents.