Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor
Actual product may vary
Thermo Scientific™

Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor

Sample well, rain, distilled/deionized, boiler feed and process waters with this adjustor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChemical Name or Material
700702pH Buffer A
700402pH Buffer B
700003pHISA Low Ionic Strength Adjustor
700902TS
also known as 700902
pH Buffer C
700001TS
also known as 700001
Low Ionic Strength pH Test Kit
Catalog number 700702
Price (USD)
342.00
Online exclusive
380.00
Save 38.00
Pack of 4
Chemical Name or Material:
pH Buffer A
Price (USD)
342.00
Online exclusive
380.00
Save 38.00
Pack of 4

Sample well water, rain water, distilled/deionized water, boiler feed water, process water and more with the Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor. It offers fast, accurate pH results in high purity and low ionic strength waters.

Features:

  • Increases measurement accuracy
  • Reduces electrode drift
  • Eliminates contamination of sample

Pure Water pH Test Kit:

  • Kit contains special low ionic strength buffers for calibration and pHISA™ adjustor additive, to increase sample ionic strength without changing its pH
  • For best results, a high quality pH electrode such as the Thermo Scientific™ Orion™ ROSS Ultra™ glass-body pH electrode, Cat. No. 8102BNUWP, is recommended
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Boiling Point100°C
Certificate of AnalysisYes
Certifications/ComplianceISO 9001
Chemical Name or MaterialpH Buffer A
ColorClear
PackagingBottle
Solubility InformationSoluble
Traceability to NISTYes
For Use With (Application)Provide fast, accurate pH results in high purity and low ionic strength waters. Recommended for samples such as well water, rain water, distilled/deionized water, boiler feed water and process water.
For Use With (Equipment)pH Meter and Electrode
GradeCertified
Physical FormLiquid
Quantity4 x 475 mL
Solution TypepH Buffer
pH6.97
Unit SizePack of 4