These products are not currently available for ordering on ThermoFisher.com. Please continue to place orders through your existing ordering channel. As part of the transition to Thermo Fisher Scientific, these products will become available through Thermo Fisher ordering channels in the coming months.
Zeta Plus™ Encapsulated System with DEL Media
Thermo Scientific™

Zeta Plus™ Encapsulated System with DEL Media

The Zeta Plus™ Encapsulated system is a single-use depth filtration solution. DEL Media incorporates lipid-specific adsorbents that help reduce lipids, surfactants and other biological contaminants from human blood plasma, serum-based products, recombinant proteins and other biological fluids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModel
7010338952E16E11ADELI
7100318960E16E01A DELP08A
7100318962E16E07A DELP08A
7010338753E16E01A DELI
Catalog number 7010338952
Price (USD)
-
Model:
E16E11ADELI
Zeta Plus™ encapsulated filters are single-use depth filters capsules for bioprocess clarification. Charged media removes particles, cells, and soluble impurities with high capacity and recovery. With manifolds in a holder, they enable flexible scaling and easy change-outs. DEL media targets lipids and surfactants, improving purity in plasma, serum, and recombinant protein processing. Available in different sizes to match process scale, supported by a range of single use scale-up capsule devices enabling testing and usage from Lab to Production Scale.
  • Efficient reduction of undesirable lipids, lipoprotein haze, and microbial and particulate contaminants.
  • Effective protection and extended life for chromatography columns and reusable membranes.
  • Flexible options for all flow and batch volume requirements.
  • Tested and optimized for pharmaceutical and biological service.
  • Safe for critical pharmaceutical, bioprocessing and biological applications.
  • Components meet USP Class VI Biological Safety Tests and requirements of CFR 21.
Specifications
For Use With (Application)Harvest And Clarification, Biopharmaceutical Manufacturing
Length (English)16 in.
Length (Metric)40.64 cm
ModelE16E11ADELI
Product LineEXT
Quantity1/Cs.
TypeEncapsulated System
Width (English)17.8 in.
Width (Metric)45.2 cm
DisposableSingle-use
Filter TypeAdsorbent Filter
Unit SizeEach