These products are not currently available for ordering on ThermoFisher.com. Please continue to place orders through your existing ordering channel. As part of the transition to Thermo Fisher Scientific, these products will become available through Thermo Fisher ordering channels in the coming months.
MicroPES™ Flat Sheet Membrane
Thermo Scientific™
MicroPES™ Flat Sheet Membrane
MicroPES™ flat sheet membranes feature an asymmetric pore structure that delivers reliable permeability and selectivity. Offered as an OEM membrane material, they empower customers to engineer, design, and manufacture filtration devices tailored to demanding industrial applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModel
7010440214W134484
7010440215W134485
7010440216W134486
7010440217W134487
7010440220W134492
7010440221W134493
7010440222W134494
7010440223W134495
7010440235W134554
7010440236W134556
7010440255W134603
7010440257W134605
7010440260W134636
7010440267W134658
7010440268W134659
7010440271W134666
7010440285W134718
7010440289W134745
7010440290W134746
7010440306W134793
7010440307W134796
7010440308W134798
7010440309W134799
7010440317W134807
7010440332W134854
7010440387W135017
7010440397W135043
7010440406W135055
7010520767W135424
7010520796W135189
7100192032W135237
7100205441W135291
7100205442W135290
7100216936W135324
7100224607W135328
7100232290W135353
7100248899W134560
7100248934W134481
7100249018W134482
7100249062W134553
7100249086W134480
7100249087W134491
7100249100W134561
7100249244W134479
7100249570W134483
Catalog number 7010440214
Price (BRL)
-
Model:
W134484
MicroPES™ Flat Sheet Membrane features an asymmetric pore structure with strong depth‐filtration characteristics for high retention and proven throughput. It can be pleated or welded using standard processes, is available in multiple pore sizes, and maintains mechanical stability during chemically enhanced backwashing due the materials broad chemical resistance (pH 2-12).
  • MicroPES™ Flat Sheet Membranes are made from polyether sulphone
  • The asymmetric pore structure of MicroPES™ Flat Sheet Membrane provides high-performance in depth filtration for a wide range of applications due to different pore sizes available
  • The MicroPES™ Flat Sheet Membrane retention layer is well protected inside the cross section
  • Good processability due to high mechanical strength
  • Can be processed with common pleating and welding processes
  • MicroPES™ Flat Sheet Membranes can be sterilized by ETO or gamma irradiation
  • MicroPES™ Flat Sheet Membranes are resistant against chemicals used in cleaning procedures, such as hydrochloric acid, citric acid, acetic acid, sodium hydroxide, or sodium hypochlorite
  • MicroPES™ Flat Sheet Membranes can be used in applications in a wide pH range from 2 to 12
  • MicroPES™ Flat Sheet Membranes passed the following toxicological tests from ISO 10993: Cytotoxicity; Skin Sensitization; Irritation or Intracutaneous Reaction; Acute Systemic Toxicity; Hemolysis; Pyrogenic Reactions; Mutagenicity in Ames-Test; Implantation Test
  • MicroPES™ Flat Sheet Membrane is EU Food Contact compliant
Specifications
For Use With (Application)Industrial Membrane Filtration
Membrane TypeFlat Sheet
ModelW134484
No. per PackVariable Pack
TypeFlat Sheet Membrane
Width (English)10 in.
Width (Metric)254 mm
Filter TypeMembrane Filter
FormatRoll
Pore Size0.1 μm
Unit SizeEach