IL-13 Antibody (21H6H5), ABfinity™ Rabbit Monoclonal

Catalog number: 701123

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Specifications

Reactivity: Mouse
Validated Application: ELISA, Western Blot
Host Species: Rabbit
SubClass (Isotype): IgG
Clonality: Monoclonal
Antibody Fragment: Whole Antibody
Gene ID: 3596
Gene Symbol: IL13
Accession Number: P35225
Label or Dye: Unconjugated
Purification: Affinity Purified
Concentration: 0.5 mg⁄ml
Form: Liquid
Mass: 100 µg
Product Size: 100 µg
Protein Family: Cytokines & Receptors
Volume (Metric): 200 µl
Research Category: Immunology
Shipping Condition: Wet Ice

Contents & storage

2 - 8°C for up to 1 month, -20°C for long storage.


Avoid repeated freezing and thawing.

Documents

Manuals & protocols

Recommended products