Rheodyne™ Sample Loops
Imagem ilustrativa. O produto pode variar
Thermo Scientific™
Rheodyne™ Sample Loops
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoVolume (Metric)For Use With (Equipment)Diameter (Metric) Inner
7020TS
conhecido também como 7020
5 μLRheodyne Injectors 7010/71250.18 mm
7024TS
conhecido também como 7024
100 μLRheodyne Injectors 7010/71250.51 mm
7025200 μLRheodyne Injectors 7010/71250.76 mm
70295 mLRheodyne Injectors 7010/71251.0 mm
7023TS
conhecido também como 7023
50 μLRheodyne Injectors 7010/71250.51 mm
Número do catálogo 7020TS
conhecido também como 7020
Preço (BRL)
-
Volume (Metric):
5 μL
For Use With (Equipment):
Rheodyne Injectors 7010/7125
Diameter (Metric) Inner:
0.18 mm
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Especificações
DescriptionFor Rheodyne 7010/7125; 5 uL; 0.18 mm ID
Diameter (English) Inner0.007 in.
Diameter (Metric) Inner0.18 mm
Volume (Metric)5 μL
For Use With (Equipment)Rheodyne Injectors 7010/7125
TypeSample Loop
Unit SizeEach