You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
▸
Shop All Products
▸
Chromatography Columns and Cartridges
▸
HPLC Columns and Cartridges
▸
HPLC Columns
▸
BetaSil™ C1 HPLC Columns
BetaSil™ C1 HPLC Columns
High surface area with high bonded phase coverage
Have Questions?
Change view
Catalog Number
Length (Metric)
Diameter (Metric)
70505-254030
250 mm
4 mm
70505-254630
250 mm
4.6 mm
2 Options
Catalog number
70505-254030
Price (BRL)
-
Request A Quote
Length (Metric):
250 mm
Diameter (Metric):
4 mm
High surface area with high bonded phase coverage
Thermo Scientific Chemistry-on 100Å silica
Specifications
Column Format
Analytical Column
Endcapped
Yes
Packing Material
Silica
Stationary Phase
C1
USP Type
L13
Carbon Load
4%
Diameter (Metric)
4 mm
Length (Metric)
250 mm
Particle Size
5 μm
Pore Size
100 Å
Product Line
Beta
Unit Size
Each