You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
New Products
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Chromatography Columns and Cartridges
▸
HPLC Columns and Cartridges
▸
HPLC Columns
▸
BetaSil™ C1 HPLC Columns
BetaSil™ C1 HPLC Columns
High surface area with high bonded phase coverage
Have Questions?
Change view
Catalog Number
Length (Metric)
Diameter (Metric)
70505-254630
250 mm
4.6 mm
70505-254030
250 mm
4 mm
2 Options
Catalog number
70505-254630
Price (KRW)
-
Length (Metric):
250 mm
Diameter (Metric):
4.6 mm
High surface area with high bonded phase coverage
Thermo Scientific Chemistry-on 100Å silica
Specifications
Column Format
Analytical Column
Endcapped
Yes
Packing Material
Silica
Stationary Phase
C1
USP Type
L13
Carbon Load
4%
Diameter (Metric)
4.6 mm
Length (Metric)
250 mm
Particle Size
5 μm
Pore Size
100 Å
Product Line
Beta
Unit Size
Each