PCR Product Pre-Sequencing Kit
PCR Product Pre-Sequencing Kit
Applied Biosystems™

PCR Product Pre-Sequencing Kit

The PCR Product Pre-Sequencing Kit uses a novel, enzymatic clean-up method to pre-treat PCR products prior to sequencing, without anyRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
70997.1.KT2000
70995.1.KT100
70996.1.KT500 Reactions
Catalog number 70997.1.KT
Price (MXN)
-
No. of Reactions:
2000
The PCR Product Pre-Sequencing Kit uses a novel, enzymatic clean-up method to pre-treat PCR products prior to sequencing, without any subsequent purification or separation steps. The two hydrolytic enzymes used in this kit, shrimp alkaline phosphatase and exonuclease I, effectively remove excess dNTPs and primers present in the final PCR product reaction mixture. The enzymes are conveniently added to an aliquot of the PCR product mixture, incubated at 37°C, and then inactivated at 80°C. The result is a cleaner PCR product, free from excess nucleotides and primers, and ready to be sequenced with standard sequencing reagents.
Specifications
DescriptionPCR Product Pre-Sequencing Kit, 2000 rxns, Recombinant, PCR Purification, Uses a Novel, Enzymatic Clean-up Method to Pre-treat PCR Products Prior to Sequencing, without Any Subsequent Purification or Separation Steps
For Use With (Application)PCR Purification
No. of Reactions2000
Product LineExoSAP-IT
Protein FormRecombinant
Quantity2000 reactions
Research CategoryGenetic Diseases
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Store at -5 to -30°C.