These products are not currently available for ordering on ThermoFisher.com. Please continue to place orders through your existing ordering channel. As part of the transition to Thermo Fisher Scientific, these products will become available through Thermo Fisher ordering channels in the coming months.
Polisher ST Membrane Adsorber
Thermo Scientific™

Polisher ST Membrane Adsorber

Polisher ST Membrane Adsorber is a single‐use AEX purifier that removes HCP, viruses, DNA, turbidity and other negatively charged contaminants in flow‐through polishing. Its hybrid design combines Q‐functional nonwoven and guanidinium chemistory to deliver high salt tolerance.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCaudalTipo de tapónCantidad
71001792290.075 gal./min.3/4 in. Sanitary Connectors1 Ea.
71001821440.001 gal./min.1 1/2 in. Sanitary Connectors1 Ea.
71002287610.001 gal./min.Luer Lock4/Cs.
71001792090.005 gal./min.3/4 in. Sanitary Connectors2/Cs.
71001792070.005 gal./min.Luer Lock2/Cs.
71001792300.037 gal./min.3/4 in. Sanitary Connectors1 Ea.
71001795600.224 gal./min.3/4 in. Sanitary Connectors1 Ea.
71001795700.506 gal./min.1 1/2 in. Sanitary Connectors1 Ea.
71002287601 mL/min.Luer Lock4/Cs.
Número de catálogo 7100179229
Precio (USD)
-
Caudal:
0.075 gal./min.
Tipo de tapón:
3/4 in. Sanitary Connectors
Cantidad:
1 Ea.
Polisher ST Membrane Adsorber is a single-use AEX purifier for flow-through polishing of biopharma streams. It reduces HCP, viruses, DNA, and other negatively charged impurities using a Q-functional nonwoven and a guanidinium membrane. The Q layer lowers turbidity and DNA, while the membrane adds capacity, strong viral clearance, and salt tolerance. Available in multiple sizes for seamless scale-up
  • The Polisher ST is single use multi-mechanism membrane adsorber which contains a Q-functional anion exchange non woven media and guanidinium functionalized membrane. This product removes residual turbidity, DNA and HCP and provides robust virus clearance for improved downstream/polishing purification.
  • Single-use technology as a cost-efficient option to increase biomass production by delivering high purity and yield in the downstream polishing unit operation in therapeutic recombinant protein process in biologically relevant operating conditions
  • Replaces downstream AEX polishing column for reduction of HCP and virus in a salt tolerant (ST) manner as a single-use solution
  • Provides DNA reduction in a salt tolerant (ST) manner compared with conventional Q-functional AEX resins
Especificaciones
Esterilizables en autoclaveAutoclavable
Peso base2.65 oz.
Volumen del lecho24.68 oz.
Material del tapónPolysulfone
Tipo de tapón3/4 in. Sanitary Connectors
ConectoresSanitary Connectors
Diámetro (imperial)9.49 in.
Diámetro (métrico)24.1 cm
Área de filtración52.7 in.²
Velocidad de filtración0.075 gal./min.
Caudal0.075 gal./min.
Para utilizar con (aplicación)Anion Exchange
JuntaFluorocarbon
Altura (imperial)3.98 in.
Altura (métrico)10.1 cm
Conexión de entrada (imperial)Sanitary Connectors
Presión máxima45 lb./in.
Cantidad1 Ea.
Temperatura máx. (imperial)104°F
Temperatura máx. (métrico)40°C
Volumen (imperial)24.68 oz.
Volumen (métrico)730 mL
Caudal0.075 gal./min.
Peso (imperial)2.65 oz.
Peso (métrico)1.2 kg
DesechablesSingle-use
Tipo de filtroSingle-use
MaterialPolysulfone
Material (membrana)Polyamide
Tamaño de poro0.8 μm
Unit SizeEach