CaptureSelect™ Biotin Anti-FVIII Conjugate
Thermo Scientific™
CaptureSelect™ Biotin Anti-FVIII Conjugate
Thermo Scientific™ CaptureSelect™ Biotin Anti-FVIII Conjugate consists of a 13 kDa recombinant single domain antibody fragment with high affinity and selectivity for both recombinant (full length and B-domain deleted) and plasma derived human Factor VIII (FVIII), through recognition of the human FVIII light chain.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
7102862500500 μg
7102862100100 μg
Número de catálogo 7102862500
Precio (MXN)
-
Cantidad:
500 μg
CaptureSelect Biotin Anti-FVIII Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant (full length and B-domain deleted) and plasma derived human Factor VIII (FVIII), through recognition of the human FVIII light chain. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
The CaptureSelect Biotin Anti-FVIII Conjugate allows you to:
  • Detect, quantitate, and characterize recombinant and plasma derived human FVIII
Applications for CaptureSelect Biotin Anti-FVIII Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Etiqueta o tinteBiotin & Other Haptens
Tipo de ligandoAnticuerpo
Cantidad500 μg
Línea de productosCaptureSelect
TipoConjugate
Unit SizeEach
Contenido y almacenamiento
Store in freezer (-5°C to -30°C).