CaptureSelect™ Biotin Anti-IgA Conjugate
Thermo Scientific™

CaptureSelect™ Biotin Anti-IgA Conjugate

Thermo Scientific™ CaptureSelect™ Biotin Anti-IgA Conjugate consists of a 14 kDa recombinant single domain antibody fragment that specifically binds to the α-chain of all human IgA antibodies (Fc domain).
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
7102882500500 μL
7102882100100 μL
카탈로그 번호 7102882500
제품 가격(KRW)
-
수량:
500 μL

CaptureSelect Biotin Anti-IgA Conjugate consists of a 14 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the α-chain of all human IgA antibodies (Fc domain). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-IgA Conjugate allows you to:

  • Detect, quantitate, and characterize all human IgA antibodies, including monomeric IgA-1 and IgA-2, dimeric IgA (secretory and non-secretory), and recombinant IgA monoclonal antibodies, as well as total IgA in human plasma and/or serum samples
  • Avoid cross-binding with other isotypes, such as IgG, IgM, IgD, and IgE, free light chains, or IgA from bovine sources such as FCS

Applications for CaptureSelect Biotin Anti-IgA Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

For Research Use Only. Not for use in diagnostic procedures.
사양
라벨 또는 염료Biotin & Other Haptens
리간드 유형Antibody
수량500 μL
타겟IgA
형태Liquid
제품라인CaptureSelect
유형Conjugate
Unit SizeEach
구성 및 보관
Store at -5°C to -30°C.