CaptureSelect™ Biotin Anti-Fibrinogen Conjugate
Thermo Scientific™
CaptureSelect™ Biotin Anti-Fibrinogen Conjugate
Thermo Scientific™ CaptureSelect™ Biotin Anti-Fibrinogen Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to human fibrinogen.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
7102912500500 μg
7102912100100 μg
Número de catálogo 7102912500
Precio (MXN)
-
Cantidad:
500 μg
CaptureSelect Biotin Anti-Fibrinogen Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to human fibrinogen. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize native and recombinant human fibrinogen.
Applications for CaptureSelect Biotin Anti-Fibrinogen Conjugate include ELISA, immunoprecipitation with streptavidin-coupled DynaBeads, MSIA Streptavidin D.A.R.T.'S, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Etiqueta o tinteBiotin & Analogs
Tipo de ligandoAnticuerpo
Cantidad500 μg
FormularioLíquido
Línea de productosCaptureSelect
TipoConjugate
Unit SizeEach
Contenido y almacenamiento
Store in freezer (-5°C to -30°C).