CaptureSelect™ Biotin Anti-GCSF Conjugate
Catalog number: 7103130500
 Related applications: Chromatography
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
CaptureSelect™ Biotin Anti-GCSF Conjugate consists of a 13 kDa camelid antibody fragment (affinity ligand) with high affinity and selectivity for both recombinant and natural sourced human granulocyte colony-stimulating factor (GCSF). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
The CaptureSelect™ Biotin Anti-GCSF Conjugate allows you to:
Detect, quantitate, and characterize recombinant and natural sourced human GCSF
Applications for CaptureSelect™ Biotin Anti-GCSF Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Ligand Type: Antibody
Product Line: CaptureSelect™
Product Size: 500 µg
Contents & storage
Store in freezer (-5 to -30°C).
Documents
Manuals & protocols