CaptureSelect™ Biotin Anti-FSH Conjugate
Thermo Scientific™
CaptureSelect™ Biotin Anti-FSH Conjugate
Thermo Scientific™ CaptureSelect™ Biotin Anti-FSH Conjugate consists of a 13 kDa recombinant single domain antibody fragment with high affinity and selectivity for both recombinant and natural sourced human follicle stimulating hormone (FSH), through recognition of an epitope specific for intact FSH only.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7103180500500 μg
7103180100100 μg
Catalog number 7103180500
Price (USD)
2,240.00
Each
Quantity:
500 μg
Price (USD)
2,240.00
Each
CaptureSelect Biotin Anti-FSH Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and natural sourced human follicle stimulating hormone (FSH), through recognition of an epitope specific for intact FSH only. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
The CaptureSelect Biotin Anti-FSH Conjugate allows you to:
  • Detect, quantitate, and characterize intact recombinant and natural sourced human FSH
  • Avoid cross-binding to dissociated free α- or β-chains of human FSH
Applications for CaptureSelect Biotin Anti-FSH Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeBiotin & Other Haptens
Ligand TypeAntibody
Quantity500 μg
FormLiquid
Product LineCaptureSelect
TypeLigand Conjugates
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).