CaptureSelect™ Biotin Anti-IgG-CH1 Conjugate
Thermo Scientific™

CaptureSelect™ Biotin Anti-IgG-CH1 Conjugate

Thermo Scientific™ CaptureSelect™ Biotin Anti-IgG-CH1 Conjugate consists of a 14 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the CH1 domain of all four human IgG subclasses.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
7103202100100 μg
7103202500500 μg
카탈로그 번호 7103202100
제품 가격(KRW)
896,000
Each
수량:
100 μg
제품 가격(KRW)
896,000
Each

CaptureSelect Biotin Anti-IgG-CH1 Conjugate consists of a 14 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the CH1 domain of all four human IgG subclasses. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-IgG-CH1 Conjugate allows you to

  • Detect, quantitate, and characterize all human IgG antibodies (subclasses 1 to 4) and Fab fragments thereof, independent of light chain type, and total IgG in human plasma and/or serum samples
  • Avoid cross-binding with other isotypes, such as IgA, IgM, IgD, and IgE, free light chains, or IgG from mouse sources or bovine sources such as FCS
  • Screen antibody-antigen interactions—biotin anti-IgG-CH1 forms a stable [low k(diss)] complex with human IgG antibodies through bivalent binding

Applications for CaptureSelect Biotin Anti-IgG-CH1 Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

For Research Use Only. Not for use in diagnostic procedures.
사양
라벨 또는 염료Biotin & Other Haptens
리간드 유형Antibody
수량100 μg
타겟IgG-CH1
형태Liquid
제품라인CaptureSelect
유형Conjugate
Unit SizeEach
구성 및 보관
Store at -5°C to -30°C.