CaptureSelect™ Biotin Anti-Insulin Conjugate

Catalog number: 7103362500

 Related applications: Chromatography

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CaptureSelect™ Biotin Anti-Insulin Conjugate consists of a 13 kDa camelid antibody fragment (affinity ligand) with high affinity and selectivity for both recombinant and natural-sourced human insulin. The affinity ligand shows similar binding reactivity towards porcine- and bovine-sourced insulin. It also binds to human proinsulin, the precursor of insulin that still contains the C-peptide. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

Applications for CaptureSelect™ Biotin Anti-Insulin Conjugate include ELISA, western blot, Gyrolab™-based immunoassays, and use of label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Ligand Type: Antibody
Product Line: CaptureSelect™
Product Size: 500 µg
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

Store at -5 to -30°C.

Documents

Manuals & protocols