CaptureSelect™ Biotin Anti-Prothrombin Conjugate
Thermo Scientific™

CaptureSelect™ Biotin Anti-Prothrombin Conjugate

Thermo Scientific™ CaptureSelect™ Biotin Anti-Prothrombin Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to human prothrombin (coagulation factor II).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7103452100100 μg
7103452500500 μg
Catalog number 7103452100
Price (CLP)
-
Quantity:
100 μg

CaptureSelect Biotin Anti-Prothrombin Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to human prothrombin (coagulation factor II). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize native and recombinant human prothrombin.

Applications for CaptureSelect Biotin Anti-Prothrombin Conjugate include ELISA, immunoprecipitation with streptavidin-coupled DynaBeads, MSIA Streptavidin D.A.R.T.'S, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeBiotin & Analogs
Ligand TypeAntibody
Quantity100 μg
FormLiquid
Product LineCaptureSelect
TypeConjugate
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).