CaptureSelect™ Biotin Anti-AAVX Conjugate
Thermo Scientific™
CaptureSelect™ Biotin Anti-AAVX Conjugate
Thermo Scientific™ CaptureSelect™ Biotin Anti-AAVX Conjugate consists of a 14 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to native and recombinant adeno-associated virus (AAV) particles of various serotypes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7103522500500 μg
7103522100100 μg
Catalog number 7103522500
Price (USD)
2,240.00
Each
Quantity:
500 μg
Price (USD)
2,240.00
Each
CaptureSelect Biotin Anti-AAVX Conjugate consists of a 14 kDa recombinant single domain antibody fragment that specifically binds with high affinity and selectivity to native and recombinant adeno-associated virus (AAV) particles of various serotypes.
The anti-AAVX affinity ligand has demonstrated binding reactivity towards a large set of different AAV serotypes including AAV1 to AAV8, and AAVrh10, indicating that an even broader range of AAV serotypes could potentially be recognized and detected. The affinity ligand is chemically conjugated to biotin via a spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
This conjugate enables the detection, quantitation, and characterization of native and recombinant AAV virus particles. Applications for CaptureSelect Biotin Anti-AAVX Conjugate include ELISA, immunoprecipitation with streptavidin-coupled Dynabeads, Gyrolab™-based immunoassays, and use with label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Ligand TypeAntibody
Quantity500 μg
Shipping ConditionDry Ice
FormLiquid
Product LineCaptureSelect
TypeConjugate
Unit SizeEach
Contents & Storage
Store at -5°C to -30°C.
Frequently asked questions (FAQs)
How many biotin molecules are there per antibody molecule in your CaptureSelect Biotin Anti-AAVX Conjugate (Cat. No. 7103522500)?
All our CaptureSelect biotin conjugates are directionally conjugated with a maximal degree of labeling of 2 biotin molecules per VHH.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.