VersaTREK™ Myco PVNA (Polymyxin B, Vancomycin, Nalidixic Acid, Amphotericin B)
VersaTREK™ Myco PVNA (Polymyxin B, Vancomycin, Nalidixic Acid, Amphotericin B)
VersaTREK™ Myco PVNA (Polymyxin B, Vancomycin, Nalidixic Acid, Amphotericin B)
Thermo Scientific™
VersaTREK™ Myco PVNA (Polymyxin B, Vancomycin, Nalidixic Acid, Amphotericin B)
Accurately detect mycobacteria in potentially contaminated specimens using Thermo Scientific™ VersaTREK™ Myco PVNA (Polymyxin B, Vancomycin, Nalidixic Acid, Amphotericin B).
Have Questions?
Catalog NumberPackaging TypeMedia TypeQuantity
7113-4225 mL EachMyco antibiotic supplement25 mL
Catalog number 7113-42
Price (MXN)
-
Packaging Type:
25 mL Each
Media Type:
Myco antibiotic supplement
Quantity:
25 mL
Accurately detect mycobacteria in potentially contaminated specimens using Thermo Scientific™ VersaTREK™ Myco PVNA (Polymyxin B, Vancomycin, Nalidixic Acid, Amphotericin B).
The VersaTREK System combines a liquid culture medium (VersaTREK Myco media), a growth supplement (Myco GS), and, for potentially contaminated species, an antibiotic supplement (Myco AS or Myco PVNA) with a detection system that automatically incubates and continuously monitors culture bottles inoculated with specimens possibly containing mycobacteria.
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Antimicrobial AgentsPolymyxin B, Vancomycin, Nalidixic Acid, Amphotericin B
DescriptionVersaTREK Myco PVNA (Polymyxin B, Vancomycin, Nalidixic Acid, Amphotericin B)
Media TypeMyco antibiotic supplement
No. per Pack5
Packaging Type25 mL Each
Quantity25 mL
Specimen TypeAll Samples (e.g. Sputum, Blood, Tissue, Urine, etc.)
Product TypeMicrobiology Media
SpeciesMycobacterium tuberculosis
Target Organism ClassMycobacteria
Test TypeMicrobial Detection, Antimicrobial Susceptibility
Unit SizeEach
Contents & Storage
2°C to 8°C