Nalgene™ Carboy Replacement Screw Closures and Gaskets
Thermo Scientific™
Nalgene™ Carboy Replacement Screw Closures and Gaskets
Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNeck FinishMaterialProduct Type
712160-083083BPolypropyleneScrew Cap
712150-1000100-400PolypropyleneScrew Cap
712155-1200120-400PolypropyleneScrew Cap
712240-105353BHDPEScrew Cap with Strap
712160-053053BPolypropyleneScrew Cap
712151-005353BHDPEScrew Closure and Gasket
712160-005353BTPESealing Gasket
712162-183083BTPESealing Gasket
712151-008383BHDPEScrew Cap
Catalog number 712160-0830
Price (USD)
68.75
Case of 2
Neck Finish:
83B
Material:
Polypropylene
Product Type:
Screw Cap
Price (USD)
68.75
Case of 2
Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service
  • Made to fit Nalgene Carboys
  • Leakproof† when used with the Nalgene bottles with the matching finish
  • † The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
    Specifications
    AutoclavableAutoclavable
    Closure Size83B
    DescriptionReplacement Closure
    DisposableSingle-use
    For Use With (Equipment)Nalgene Large Carboys and Bottles
    Neck Finish83B
    No. per Case2
    No. per Pack2
    Product TypeScrew Cap
    ColorWhite
    MaterialPolypropylene
    SterilityNon-sterile
    Unit SizeCase of 2