You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
首頁
▸
瀏覽所有產品
▸
Burettes
▸
General Purpose Burets
▸
Replacement Stopcock Assembly for Nalgene™ Acrylic Buret
Replacement Stopcock Assembly for Nalgene™ Acrylic Buret
Accurately perform applications with Thermo Scientific™ Nalgene™ Acrylic Burets with PMP Tip, TFE Stopcock and Plug, which meet ASTM E287 standards.
Have Questions?
產品號碼
For Use With (Equipment)
713650-1025
25 mL Acrylic Nalgene Break-Resistant Burets
產品號碼
713650-1025
價格 (HKD)
1,920.00
Each
新增至購物車
For Use With (Equipment):
25 mL Acrylic Nalgene Break-Resistant Burets
價格 (HKD)
1,920.00
Each
新增至購物車
This is a replacement part for Nalgene™ Acrylic Buret 3650-0025.
This is a replacement part for Nalgene™ Acrylic Buret 3650-0025.
規格
For Use With (Equipment)
25 mL Acrylic Nalgene Break-Resistant Burets
Length (Metric)
25 mm
Material
PTFE
Type
Replacement Stopcock Assembly
Unit Size
Each