VersaTREK™ Connectors
VersaTREK™ Connectors
VersaTREK™ Connectors
VersaTREK™ Connectors
VersaTREK™ Connectors
Thermo Scientific™
VersaTREK™ Connectors
Facilitate organism detection by linking culture bottle pressure changes to the Thermo Scientific™ VersaTREK™ instrument sensor while preventing gas build up in the bottles.
Have Questions?
Catalog NumberPackaging Type
7150-4450/Box
Catalog number 7150-44
Price (MXN)
-
Packaging Type:
50/Box
Facilitate organism detection by linking culture bottle pressure changes to the Thermo Scientific™ VersaTREK™ instrument sensor using Thermo Scientific VersaTREK Connectors. The recessed probe also prevents dangerous gas build up within the Thermo Scientific™ VersaTREK™ REDOX™ or Thermo Scientific VersaTREK Myco bottle.
Meet the demands of both high and low volume laboratories with the Thermo Scientific VersaTREK System, which comes with a full line of ancillary and accessory products. With four FDA-cleared tests and a simple, two-bottle media system, the VersaTREK System provides maximum value with faster results and fewer limitations.
  • Only system capable of detecting any gas produced or consumed by organisms, allowing detection of a wider range of both common and fastidious organisms
  • Simplified workflow includes easy scan function for bottle entry, intuitive icons to reduce training time, and no daily QC requirement for maximum efficiency
  • Highly-enriched Thermo Scientific VersaTREK REDOX media supports a wide range of organisms, facilitating better recovery of fastidious organisms
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionVersaTREK Connectors
No. per Pack50
Packaging Type50/Box
Specimen TypeVarious
Product TypeMicrobiology Media
SpeciesM. avium ssp. paratuberculosis
Test TypeMicrobial Detection
Unit SizeEach