Thermo Scientific™
HA Tag Rabbit anti-Tag, Polyclonal, Invitrogen™
Related applications:
Anatomical Pathology
Maintain Thermo Scientific™ HM 430 and HM 450 Sliding Microtomes with Thermo Scientific HM 430, HM 450 and 440 E Sliding Microtomes Accessories.
Back to top
Epitope tags provide a method to localize gene products in a variety of cell types; study the topology of proteins and protein complexes; identify associated proteins; and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. An HA tag is frequently incorporated into recombinant proteins for a variety of purposes. An anti-HA antibody can then be used to detect the protein when doing studies with transfected cells.