Nalgene™ Staining Box
Nalgene™ Staining Box
Nalgene™ Staining Box
Nalgene™ Staining Box
Thermo Scientific™
Nalgene™ Staining Box
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods with these staining boxes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
715705-0001Staining Box Plug
5705-1010Gel Staining Box
Catalog number 715705-0001
Price (CLP)
-
Type:
Staining Box Plug
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods. Thermo Scientific™ Nalgene™ Staining Box is equipped with a bottom drain allowing you to change solution without disturbing the box's other fragile contents. Box holds 4.8 in. x 4.8 in (189 mm x 189 mm) gel along with 500mL of liquid.
  • Polymethylpentene box and polypropylene copolymer cover
  • Useful over a -70° to +70°C (-94° to +158°F) temperature range
  • Good chemical compatibility with acids, bases and select organic solvents for a versatile range of protocols
  • LDPE plug in bottom for easy drainage of liquid with minimal handling of gel and provides a leakproof seal
  • Plug won't interfere with cover or contents of boxes on a benchtop or shaker
  • Stackable, with a handle on each end for safe, easy carrying
  • Microwavable and easy to clean
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene  products are packaged.
Specifications
DescriptionReplacement Plug for 5705-1010 and -2020
No. per Case12
No. per Pack12
TypeStaining Box Plug
Unit SizeCase of 12