Thermo Scientific™

HM 650V Vibrating Blade Microtome Accessories

Related applications:

Anatomical Pathology

Use these Thermo Scientific™ HM 650V Vibrating Blade Microtome Accessories with the Thermo Scientific™ HM650V Vibrating Blade Microtome.
Back to top
Standard buffer tray:
  • Hold stages in place using a magnet
  • Fill tray with all water-based buffers and solutions
Cooling element for buffer solution:
  • Use pre-cooled in the standard buffer tray for cooling tissue
  • Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
Large-field magnifier:
  • Magnification: 2.5x
  • Ability to move, swivel, and displace for optimal viewing position
  • Adapter and illumination included
StemiScope™ with optics and mount:
  • Eyepieces: 10x
  • Supplementary lens: 0.63x
  • Zoom magnifying range: 4.1 to 31.5x
  • Microscope adapter for angled view of specimen
  • Horizontal view and vertical drive and swivel mechanism
Swan-neck light guide:
  • 3.5mm fiber bundle
  • 50mm long swan-neck light guide
  • 750mm long flexible light guide
  • High light intensity
  • Adapter for the KL200 (760317) included