Product Image
Thermo Scientific™

Petri Dish Rack Post

Post Polycarbonate for use with 5920/21-0060 5922-0024 and 5924-0022 petri dish rack
Have Questions?
Catalog NumberPositions
715920-0650Post
Catalog number 715920-0650
Price (BRL)
-
Positions:
Post

Post Polycarbonate for use with 5920/21-0060 5922-0024 and 5924-0022 petri dish rack

  • White Polycarbonate petri dish rack post has smooth, rounded corners, finger-grip handle and Rubber feet which make carrying up to 54 plastic-round dishes (100 x 15 mm) or 42 glass-round dishes (100 x 15 mm) safe and secure
  • Petri dish rack can be decontaminated by heat sterilization
  • Simple assembly required
  • Material: Polycarbonate
  • Color: White
  • Size: 6.5 in.
Specifications
For Use With (Equipment)Petri Dish Rack Post
Size6.5 in.
AutoclavableAutoclavable
ColorWhite
MaterialPolycarbonate
PositionsPost
Product TypePetri Dish Rack
SterilitySterile
Unit SizePack of 4