Product Image
Invitrogen™

Anti-Human Foxp3 Staining Set PE, eBioscience™

This Anti-Human Foxp3 Staining Set contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3 cells. The자세히 알아보기
Have Questions?
카탈로그 번호수량
72-5774-401 kit
카탈로그 번호 72-5774-40
제품 가격(KRW)
554,000
온라인 행사
Ends: 30-Sep-2026
651,000
할인액 97,000 (15%)
Each
수량:
1 kit
제품 가격(KRW)
554,000
온라인 행사
Ends: 30-Sep-2026
651,000
할인액 97,000 (15%)
Each
This Anti-Human Foxp3 Staining Set contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3 cells. The 236A/E7 monoclonal antibody reacts with human Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2. Foxp3 is a 49-55 kDa protein and a member of the forkhead/winged-helix family of transcription factors. It was identified as the gene responsible for the X-linked lymphoproliferative disease observed in scurfy (sf) mice and in the human disorder, X-linked autoimmunity-allergic dysregulation syndrome (XLAAD). Constitutive expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

The 236A/E7 antibody has been shown to crossreact with rhesus macaque, sooty mangabey, and cynomolgus Foxp3 236A/E7 recognizes a different epitope of Foxp3 than PCH101 and 150D/E4.

Isotype
IgG1

Laser
Blue Laser

Emit
575 nm

Excite
488 nm

Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.
사양
항원Foxp3
클론236A/E7
유전자 별칭Forkhead Box P3, Scurfin, JM2
일차 또는 이차Primary
제품라인eBioscience
애플리케이션Flow Cytometry
분류Monoclonal
콘주게이트PE
숙주 종Mouse
IsotypeIgG1
수량1 kit
인허가 상태RUO
타겟 종Cynomolgus, Human, Macaque, Monkey, Non-human Primate, Rhesus Monkey
Unit SizeEach
구성 및 보관
Store at 2-8°C. Light sensitive material. Use within 6 months of opening or by date indicated on the bottle.