Product Image
Invitrogen™

Anti-Human Foxp3 Staining Set PE, eBioscience™

This Anti-Human Foxp3 Staining Set contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3 cells.
Have Questions?
Catalog NumberQuantity
72-5776-401 Kit
Catalog number 72-5776-40
Price (BRL)
3.632,90
Each
Quantity:
1 Kit
Price (BRL)
3.632,90
Each

This Anti-Human Foxp3 Staining Set contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3 cells. The PCH101 monoclonal antibody reacts with the amino terminus of human Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2. Foxp3 is a 49-55 kDa protein and a member of the forkhead/winged-helix family of transcription factors. It was identified as the gene responsible for the X-linked lymphoproliferative disease observed in scurfy (sf) mice and in the human disorder, X-linked autoimmunity-allergic dysregulation syndrome (XLAAD). Constitutive expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

The PCH101 antibody crossreacts with rhesus, chimpanzee, and cynomolgus Foxp3 PCH101 recognizes a different epitope of Foxp3 than clones 236A/E7 and 150D/E4.

  • Isotype: IgG2a, kappa
  • Laser: Blue Laser
  • Excitation/Emission: 488/575 nm
  • Reported Applications: Intracellular Staining followed by Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Cell TypeFoxp3
Excitation/Emission488/575 nm
Product LineeBioscience
Product TypeAnti-Human Foxp3 Staining Set
IsotypeIgG2a Kappa
Quantity1 Kit
Unit SizeEach