Chromeleon™ Chromatography Data System eLearning courses
Thermo Scientific™

Chromeleon™ Chromatography Data System eLearning courses

The new Chromeleon CDS eLearning on-demand basic user training and user administrator training courses are designed to provide an accessible and consistent experience to all learners allowing them to boost productivity and optimize budget allocation through a standardized learning experience
Have Questions?
變更視圖buttonViewtableView
產品號碼Course Topic
7202.0006Administrator Training
7202.0005Basic Operations
產品號碼 7202.0006
價格 (HKD)
-
Course Topic:
Administrator Training

The new Chromeleon CDS eLearning on-demand courses are designed to provide an accessible and consistent experience to all learners allowing them to boost productivity and optimize budget allocation through a standardized learning experience. You will find a series of eLearning courses which can cover different topics such as Chromeleon CDS basic user training and user administrator training.

Whether you are a laboratory user or a system administrator, we've got you covered. Our new Chromeleon CDS eLearning on-demand courses are designed to provide an accessible and consistent experience to all learners.

Boost productivity and optimize budget allocation through a standardized learning experience. Our eLearning program is built for:

  • Accessibility: Access all modules online anywhere at any time.
  • Flexibility: Access the eLearning material at your own pace, and it's always available for a refresher or reinforcement training, further increasing the value of your subscription.
  • Cost-Effectiveness: Eliminate the need for physical training spaces or travel expenses associated with attending in-person classes.
  • Consistency: Unify your teams across the world with the same learning experience.
  • Partnership: Amplify your learning experience by joining monthly Q&A (Basic user training only) sessions with our CDS experts.
  • You: Combine our modules to support your needs for your business as a user or a system administrator.

We offer the following Chromeleon CDS On-Demand courses:

Chromeleon CDS basic user training is a self-paced eLearning course designed for chromatographers new to Chromeleon CDS or perhaps those that need a refresher on the basics of the product. The course covers all basic aspects of Chromeleon CDS, allowing a new user to perform all steps required to operate their chromatography system from starting the software all the way to producing results. Prior experience with Chromeleon software is not required, but learners should have a working knowledge of the fundamentals of chromatography, as well as familiarity with Microsoft Windows™.

Amongst the topics covered in this eLearning series are:

  • How to Launch Chromeleon CDS software
  • Introduction to the Console
  • Introduction to the Console, Navigation Panes and eWorkflows
  • Navigation Panes
  • eWorkflows
  • How to use DataVaults
  • How to use the Help features
  • How to import data
  • How to create and run sequences
  • How to create basic instrument methods
  • How to create processing methods
  • How to create and use detection parameters
  • How to use the Cobra Wizard
  • Using Calibration Modes in Chromeleon CDS software
  • Printing and Exporting

Chromeleon CDS user administrator training is targeted to those who will be responsible for the configuration and administration of a Chromeleon Enterprise environment. The topics range from an introduction of the overall architecture to a granular look at the policies, privileges and management functionality in the Chromeleon Administration Console.

Amongst the topics covered in this eLearning series are:

  • Introduction to Chromeleon Administration
  • Introduction to Chromeleon Software Architecture
  • Introduction to Chromeleon Network Architecture
  • Introduction to Chromeleon Administration Console
  • Chromeleon Administration Console License Manager
  • License Manager
  • Scheduler
  • Global Policies
  • User Database
  • eWorkflow Tags
  • Audit Trail
  • Local Machine
  • Services Manager
規格
Access Availablity12 months upon end-user first access to the course
Course TopicAdministrator Training
DescriptionChromeleon CDS user administrator training/eLearning course
FormatSelf-paced eLearning
LanguagesEnglish
TechniqueChromeleon CDS
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration2 to 4 hr.
For Use With (Equipment)Chromeleon 7.2 SR5 CDS and higher
Product LineThermo Scientific
Product TypeSoftware
Target AudienceUser seeking to learn about specific instruments/software features
Training LocationDigital access through Online Learning Management System
Unit SizeEach

常見問答集 (常見問題)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.